missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Wide-Form Porcelain Crucibles

Withstand temperatures up to 1150°C (2102°F)

Manufacturer:  Fisherbrand™ J136

 View more versions of this product

Catalog No. FB965J



 

Alert:

Heating and cooling rates should not exceed 200°C per hour and maximum use temp of 1100°C


Specifications

Description

Crucible, Porcelain; Fisherbrand; Silico; High form; w/o Cover; Top O.D x Height x Wall Thickness: 41 x 26 x 2mm; 17g; Case wt.: 1.9kg; Case size: 300L x 180Bx 260 in. H; Capac.: 17mL; 6/Pk.; 8 Pk./Cs.

  • Glazed inside and out except for outside bottom surface and rim

Description & Specifications

Specifications

1.61 in.
17g
Crucible
Porcelain
26mm
1150°C
2102°F
0.57 oz.
0.04 lb.
41mm
17mL
0.57 oz., 17mL
2mm
0.078 in.
Glazed
1.02 in.
Accessories