Please login to your online account to display your discounted pricing
Fisherbrand™ White Vinyl-Coated Test Tube Racks with Legs
Resist chemical attack from dilute mineral acids and alkalis
$46.46 - $46.46
Specifications
Autoclavable Yes
Material Epoxy Coated Heavy-gauge Steel Wire
Type Rack, Tube
View More Specs
Products
Catalog Number Mfr. No. Diameter (Metric) Cavity Dimensions (L x W x H) Holds Positions Price Quantity    
14-790-4
fisherbrand™
187854834
16mm 9.5 x 3.37 x 2.5 in. (24.1 x 8.6 x 6.4cm) Each for $46.46
Case of 12 Each for $557.52
14-790-3
fisherbrand™
187859034
13mm 9.62 x 4 x 2.25 in. (24.4 x 10 x 5.7cm) Each for $46.46
Case of 12 Each for $557.52
14-790-5
fisherbrand™
187854034
22mm 10.25 x 4.25 x 3.25 in. (26 x 11 x 8.3cm) Each for $46.46
Case of 12 Each for $557.52
14-802-8
fisherbrand™
187857234
16mm 9.25 x 4.75 x 2.75 in. (23.5 x 12 x 7cm) Each for $46.46
Case of 12 Each for $557.52
Description & Specifications
Specifications
Autoclavable Yes
Material Epoxy Coated Heavy-gauge Steel Wire
Type Rack, Tube
  • Epoxy-coated, heavy-gauge steel wire racks with three decks and legs
  • Reduce noise; extremely impact resistant
  • Usable at temperatures to 100°C (212°F)
  • Autoclavable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.