Please login to your online account to display your discounted pricing

Fisherbrand™ White Polypropylene Caps with PTFE Liners


Economical alternative to Thermoset and Phenolic PTFE lined caps. Fisherbrand™ White Polypropylene Caps have PTFE facing backed by F217 liners.

Manufacturer: fisherbrand™  263818L

 View more versions of this product

Catalog No. 02-912-075

  • / Pack of 100 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Sterile Yes
Product Type PE Foam/PTFE Lined Cap
Closure Color White
Closure Material Polypropylene
Closure Size 48/400
Closure Type Screw
Description Screw cap
Liner Material PE Foam/PTFE
Packaging Type Packed in 5 bags of 100
Quantity 500EA/Cs.
Shape Round
Certifications/Compliance ISO 9001
Closure Liner PTFE/Polyethylene
Color White
For Use With Bottle
Material Polypropylene
Neck Finish 48-400
No. per Case 100/Pk.

  • Features F217 liners which consists of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE)
  • PTFE/F217 liners are resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference