Please login to your online account to display your discounted pricing

Fisherbrand™ White Polypropylene Caps with PTFE Liners


Economical alternative to Thermoset and Phenolic PTFE lined caps. Fisherbrand™ White Polypropylene Caps have PTFE facing backed by F217 liners.

Manufacturer: fisherbrand™  263817L

 View more versions of this product

Catalog No. 02-912-074

  • / Case of 500 Each
 

Description & Specifications

Specifications

Closure Size 45/400
Closure Material Polypropylene
Shape Round
Sterile Yes
Liner Material PE Foam/PTFE
Closure Type Screw
Closure Color White
Description Screw cap
Product Type PE Foam/PTFE Lined Cap
Quantity 500EA/Cs.
Packaging Type Packed in 5 bags of 100
Color White
Material Polypropylene
Certifications/Compliance ISO 9001
Closure Liner PTFE/Polyethylene
No. per Case 500/Cs.
For Use With Bottle
Neck Finish 45-400

  • Features F217 liners which consists of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE)
  • PTFE/F217 liners are resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference