Please login to your online account to display your discounted pricing

Fisherbrand™ White Polypropylene Caps with PTFE Liners


Economical alternative to Thermoset and Phenolic PTFE lined caps. Fisherbrand™ White Polypropylene Caps have PTFE facing backed by F217 liners.

Manufacturer: fisherbrand™  263816L

 View more versions of this product

Catalog No. 02-912-073

  • / Case of 500 Each
 

Description & Specifications

Specifications

Closure Color White
Type PE Foam/PTFE Lined Cap
Closure Size 43/400
Neck Finish 43-400
Shape Round
For Use With (Equipment) Bottle
Liner Material PE Foam/PTFE
Sterility Sterile
Packaging Packed in 5 bags of 100
Closure Type Screw
Quantity 500EA/Cs.
Description Screw cap
Closure Material Polypropylene
Material Polypropylene
Certifications/Compliance ISO 9001
Color White
No. per Case 500/Cs.
Closure Liner PTFE/Polyethylene

  • Features F217 liners which consists of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE)
  • PTFE/F217 liners are resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference