Fisherbrand™ White Polypropylene Caps with PTFE Liners
Economical alternative to Thermoset and Phenolic PTFE lined caps. Fisherbrand™ White Polypropylene Caps have PTFE facing backed by F217 liners.
Manufacturer:  Fisherbrand™ 263812L
 View more versions of this product
Catalog No. 02-912-069
Specifications
Description
Cap, Screw; Fisherbrand; PP; PTFE faced PE-lined; For analytical lab samples, high-purity chemicals, strong acids and solvents; 24mm-400 screw cap size; 100/pk.
  • Features F217 liners which consists of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE)
  • PTFE/F217 liners are resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference
Description & Specifications
Specifications
ISO 9001
PTFE/Polyethylene
Screw cap
100/Pk.
500EA/Cs.
Cap, PE Foam/PTFE Lined
White
PE Foam/PTFE
Round
Polypropylene
White
Polypropylene
Packed in 5 bags of 100
Sterile
Bottle
24/400
24-400
Screw