Please login to your online account to display your discounted pricing

Fisherbrand™ White Polypropylene Caps with PTFE Liner

For use with analytical lab samples, high-purity chemicals, strong acids and solvents. Fisherbrand™ White Polypropylene Caps with PTFE Liners are an economical alternative to Thermoset and Phenolic PTFE lined caps.

$295.97 - $314.50

Specifications

Certifications/Compliance ISO 9001
Closure Liner PTFE/Polyethylene
Color White
For Use With Bottle
View More Specs
Products
Catalog Number Mfr. No. No. per Case Neck Finish Price Quantity    

02-912-065

 
fisherbrand™
263808L
500/Cs. 15-425 Case of 500 Each for $314.50

02-912-068

 
fisherbrand™
263811L
500/Cs. 22-400 Case of 500 Each for $295.97
Description & Specifications

Specifications

Certifications/Compliance ISO 9001
Closure Liner PTFE/Polyethylene
Color White
For Use With Bottle

  • Ideal for environmental samples, pharmaceuticals and diagnostic reagents
  • Screw cap sizes for narrow- and wide-mouth bottles

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.