Please login to your online account to display your discounted pricing

Fisherbrand™ White Polypropylene Caps with PTFE Liner


For use with analytical lab samples, high-purity chemicals, strong acids and solvents. Fisherbrand™ White Polypropylene Caps with PTFE Liners are an economical alternative to Thermoset and Phenolic PTFE lined caps.

Manufacturer: fisherbrand™  263808L

 View more versions of this product

Catalog No. 02-912-065

  • / Case of 500 Each
 

Description & Specifications

Specifications

Certifications/Compliance ISO 9001
Closure Liner PTFE/Polyethylene
Color White
Material Polypropylene
No. per Case 500/Cs.
For Use With (Equipment) Bottle
Neck Finish 15-425

  • Ideal for environmental samples, pharmaceuticals and diagnostic reagents
  • Screw cap sizes for narrow- and wide-mouth bottles