Please login to your online account to display your discounted pricing

Fisherbrand™ White Polypropylene Caps with PTFE Liners


Economical alternative to Thermoset and Phenolic PTFE lined caps. Fisherbrand™ White Polypropylene Caps have PTFE facing backed by F217 liners.

Manufacturer: fisherbrand™  263821L

 View more versions of this product

Catalog No. 02-912-085

  • / Case of 288 Each
 

Description & Specifications

Specifications

For Use With (Equipment) Bottle
Liner Material PTFE faced PE
Neck Finish 63-400
Closure Type Screw cap
Closure Size 63/400
Material Plastic
Closure Material Plastic
Description Screw cap
Quantity 288/PK.
Color White
Certifications/Compliance ISO 9001
No. per Case 288/Cs.
Closure Liner PTFE/Polyethylene

  • Features F217 liners which consists of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE)
  • PTFE/F217 liners are resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference