Please login to your online account to display your discounted pricing

Fisherbrand™ White Polypropylene Caps with PTFE Liners


Economical alternative to Thermoset and Phenolic PTFE lined caps. Fisherbrand™ White Polypropylene Caps have PTFE facing backed by F217 liners.

Manufacturer: fisherbrand™  W014503

 View more versions of this product

Catalog No. 02-912-076

  • / Pack of 100 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Closure Color White
Type PE Foam/PTFE Lined Cap
Packaging Packed in 5 bags of 100
For Use With (Equipment) Bottle
Neck Finish 53-400
Shape Round
Closure Size 53/400
Closure Type Screw
Liner Material PE Foam/PTFE
Sterility Sterile
Closure Material Polypropylene
Description Screw cap
Quantity 500EA/Cs.
Certifications/Compliance ISO 9001
Material Polypropylene
Color White
No. per Case 100/Pk.
Closure Liner PTFE/Polyethylene

  • Features F217 liners which consists of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE)
  • PTFE/F217 liners are resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference