Please login to your online account to display your discounted pricing

Fisherbrand™ White Plastic Screw Bottle Caps

Manufacturer: fisherbrand™  W014492

 View all options for this product

Catalog No. 02-912-067


Description & Specifications


Certifications/Compliance ISO 9001
Closure Color White
Closure Material Polypropylene
Closure Liner PTFE/Polyethylene
Closure Size 20/400
Closure Type Screw
Color White
Description Screw cap
For Use With Bottle
Material Polypropylene
Liner Material PE Foam/PTFE
Neck Finish 20-400
No. per Case 100/Pk.
Packaging Type Packed in 5 bags of 100
Quantity 500EA/Cs.
Shape Round
Sterile Yes
Product Type PE Foam/PTFE Lined Cap

1mm polyvinyl film bonded to 1mm HDPE on a #30 white pulp paper backing. Superior to plain pulp paper because it provides excellent moisture barrier. For Fisherbrand™ Boston round and wide mouth packer bottles. General purpose suitable for a wide range of applications. Good chemical resistance suitable for mild acids, alkalis, solvents, alcohols, oils and aqueous products; poor for active hydrocarbons and bleaches.