Please login to your online account to display your discounted pricing
Fisherbrand™ White Line Flasks with standard taper Stoppers
Single graduation ring blasted on the neck
Manufacturer: fisherbrand™  10205J
 View more versions of this product
Catalog No. 10-205J
  • / Case of 6 Each
Description & Specifications
Specifications
Stopper Number 13
Material Borosilicate Glass
Graduated Yes
Stopper Material Glass
Capacity (Metric) 150mL
Calibration To contain
Total Count 1.0