Fisherbrand™ Wash Bottles with Adjustable Sprayer
Handle hot/cold, aqueous/solvent-based solutions
Manufacturer:  Fisherbrand™ CON3345
 View more versions of this product
Catalog No. 03-405A
Description
Description
Handle hot/cold, aqueous/solvent-based solutions using Fisherbrand™ Wash Bottles with Adjustable Sprayer.
Fully adjustable spray pattern: “fine mist” for TLC plates; “needle” for rinsing glassware; “big stream” for powerful jet needed to wash filters. 
  • Bottle is made of high-density polyethylene
  • Sprayer is polypropylene
  • Trigger allows one-handed, controlled delivery with light pressure
  • Engraved markings show how much fluid is left
  • Squat shape takes up little bench space and ensures a low center of gravity
Specifications
Specifications
8 oz., 240mL
8 oz.
HDPE, HDPE
Squat
240mL
Polypropylene adjustable sprayer
Non-sterile
Documents