Fisherbrand™ Viscometer Holders
For use with both calibrated and uncalibrated Cannon™-Fenske Routine, Cannon-Fenske opaque, Cannon-Manning semimicro and Cannon-Manning vacuum viscometers.
Manufacturer:  Fisherbrand™ 9726M50
 View more versions of this product
Catalog No. 13-617-6
Description
Description
For use with both calibrated and uncalibrated Cannon™-Fenske Routine, Cannon-Fenske opaque, Cannon-Manning semimicro and Cannon-Manning vacuum viscometers.
  • Available in either molded plastic or neoprene rubber to fit holes with a 2 in. diameter (5.1cm)
  • Plastic holders feature spring clip to hold viscometer securely
Specifications
Specifications
Molded Plastic
Calibrated and uncalibrated Cannon™ fenske routine, Cannon™ fenske opaque and Cannon™ manning semimicro viscometers
Viscometer Holder
Documents