Please login to your online account to display your discounted pricing

Fisherbrand™ VersaClosure™ Tube Closures


Universal cap closure

Manufacturer: fisherbrand™  A20013FIS

 View more versions of this product

Catalog No. 02-707-18

  • / Pack of 1000 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

Autoclavable No
Color Red
Material LDPE
For Use With (Equipment) 16, 13, 12 and 10mm blood collection, glass and plastic test tube
Sterile No