Fisherbrand™ Vacuum Filter Holders: Silicone Rubber Stoppers
For use with Fisherbrand Vacuum Filter Holders
Manufacturer:  Fisherbrand™ 097539
 View more versions of this product
Catalog No. 09-753-9
Specifications
Description
Stopper; Accessory for vacuum filter holder; For Use with Cat. No.: 09-753A, 09-753C, 09-753E, 09-753G; Description: No. 8B; adapts stem to standard manifold
  • Constructed of durable silicone
Description & Specifications
Specifications
Stopper, No. 8b, Adapts to Manifold
Silicone