Please login to your online account to display your discounted pricing
Fisherbrand™ Urine Sedimentation System
Designed to produce the same results as the most popular style urine sedimentation system using the same test procedure
$85.21 - $296.46
Specifications
For Use With (Application) Designed to produce the same results as the most popular style Urine sedimentation system using the same test procedure
For Use With (Equipment) Urine sedimentation system
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    
14-377-260
fisherbrand™
14377260
Trans IM tube with cap Case of 500 Each for $296.46
14-375-238
fisherbrand™
14375238
Tube cap Case of 1000 Each for $85.21
14-375-560
fisherbrand™
BMPUR590
Bulb decanting pipette Pack of 500 Each for $180.21
Case of 10 Pack for $1,802.10
14-375-561
fisherbrand™
14375561
Tube Case of 500 Each for $145.21
Description & Specifications
Specifications
For Use With (Application) Designed to produce the same results as the most popular style Urine sedimentation system using the same test procedure
For Use With (Equipment) Urine sedimentation system
  • 15mL graduated urine tube in economical or break-resistant polystyrene
  • Decanting pipet allows supernatant to be decanted off; leaves 1mL of urine-containing sediment in the bottom of the tube
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.