missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Upholstered Saddle Stool and Chair

Give your staff ergonomic relief for standing positions with a Fisherbrand Upholstered Saddle Stool and Chair.

$1,227.67 - $1,227.67

Specifications

Adjustable (Height) Yes
Material Black Upholstery
For Use With (Application) Ergonomic relief for standing position
Height (English) 25 to 34.5 in.
Height (English) Base 25 to 34.5 in.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Model Product Type Price Quantity  
Catalog Number Mfr. No. Model Product Type Price Quantity  
14-359-202 Fisherbrand™
91830BUP
Chair Saddle Chair Each for $1,227.67
Specifications

Description

  • Durable saddle seat
  • Adjustable height from 25 to 34.5 in. (63.5 to 87.6cm)
  • Upholstered for greater comfort

Description & Specifications

Specifications

Yes
Ergonomic relief for standing position
25 to 34.5 in.
63.5 to 87.6cm
Pneumatic
Black Upholstery
25 to 34.5 in.
25 to 34.5 in.
63.5 to 87.6cm
Adjustable