Please login to your online account to display your discounted pricing

Fisherbrand™ Unlined Plastic Caps

Unlined caps

$195.00 - $308.70

Specifications

Certifications/Compliance ISO 9001
Color Black
Material Plastic
For Use With Bottle
View More Specs
Products
Catalog Number Mfr. No. No. per Case Neck Finish Price Quantity    

02-912-334

 
fisherbrand™
W014538
1100 Cs. 58-400 Case of 1100 Each for $308.70

02-911-847

 
fisherbrand™
W014537
288 Cs. 58-400 Case of 288 Each for $195.00
Description & Specifications

Specifications

Certifications/Compliance ISO 9001
Color Black
Material Plastic
For Use With Bottle

  • Available in various sizes and packs

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.