Please login to your online account to display your discounted pricing

Fisherbrand™ Unlined Plastic Caps


Unlined caps

Manufacturer: fisherbrand™  W014537

 View more versions of this product

Catalog No. 02-911-847

  • / Case of 288 Each
 

Description & Specifications

Specifications

For Use With Bottle
Material Plastic
Certifications/Compliance ISO 9001
Color Black
Neck Finish 58-400
No. per Case 288 Cs.