Please login to your online account to display your discounted pricing

Fisherbrand™ Unlined Plastic Caps


Unlined caps

Manufacturer: fisherbrand™  W014538

 View more versions of this product

Catalog No. 02-912-334

  • / Case of 1100 Each
 

Description & Specifications

Specifications

Certifications/Compliance ISO 9001
Color Black
Material Plastic
No. per Case 1100 Cs.
For Use With (Equipment) Bottle
Neck Finish 58-400