Fisherbrand™ Universal All Purpose Dark Green Absorbent Rolls
Designed for high wear resistance and ideal for foot traffic and walkways
Manufacturer:  Fisherbrand™ FSCRECALL30150M
 View more versions of this product
Catalog No. 19140930
Description
Description
Designed for high wear resistance and ideal for foot traffic and walkways
  • General Purpose absorbents are made with MAXX™ Technology for superior absorbency and greater strength without the added weight
  • Made of all-purpose polypropylene suitable for most industrial applications
Specifications
Specifications
Heavy
49 gal.
45.7m
Dark Green Universal - All Purpose Absorbent Roll
30 in.
45.7m x 76.2cm (150 ft. x 30 in.)
Dark green
185.4L
150 ft.
Polypropylene
76.2cm
Documents