Fisherbrand™ Universal - All Purpose Dark Green Absorbent Pads
Designed for high wear resistance and ideal for foot traffic and walkways
$80.98 - $88.30
Specifications
Color Dark green
Material Polypropylene
Product Type Dark Green Universal - All Purpose Absorbent Pad
Capacity (Metric) 98.4L
Dimensions (L x W) 48.2 x 38.1cm (19 x 15 in.)
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Absorbancy Quantity Price Quantity  
Catalog Number Mfr. No. Absorbancy Quantity Price Quantity  
19-140-944 Fisherbrand™
FSCRECALL1519L200
Light, 9 gl/gs 200/Cs. Case of 200 Each for $88.30
19-140-929 Fisherbrand™
FSCRECALL1519M100
Heavy, 12 gl/gs 100/Cs. Case of 100 Each for $80.98
Specifications
Description
Compliance
OSHA 29 CFR 1910.22(a)(2); EPA 40 CFR 112.7(c)(1)(vii); OSHA 29 CFR 1910,120(j)(1)(vii).
Description & Specifications
Specifications
Dark green
Dark Green Universal - All Purpose Absorbent Pad
48.2 x 38.1cm (19 x 15 in.)
Polypropylene
98.4L