Fisherbrand™ Ultra Flex Clamping Systems
Unique flexible arms that are extremely versatile and can be placed in virtually any position or angle.
Manufacturer:  Fisherbrand™ 916283
 View more versions of this product
Catalog No. 02-217-013
Description
Description
  • Ultra Flex comes in three different systems: base plate, lab-frame connector, and bench clamp
  • Each system includes a 2-Prong Clamp Head, 3-Prong Clamp Head, Spring Clamp Head, nickel-plated flex arm in 305mm (12 in.) or 457mm (18 in.) length, and utility wrench
  • 2-prong and 3-prong clamp heads are also supplied with non-slip vinyl sleeves and, for temperatures above 100°C, fiberglass covers
Specifications
Specifications
2.5 in.
Two Prong Clamp Head
7.49cm
6.35cm
2.95 in.
Ultra flex clamping system components
Documents