missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ U.S. Standard Stainless Steel Test Sieves, 12 in. dia. × 3.25 in.D

Stainless steel. Aperture sizes from 5.00 in. opening to 500 mesh.

Manufacturer:  Fisherbrand™ 6658

 View more versions of this product

Catalog No. 04-884-1AA


 

Description

Description

Stainless steel. Aperture sizes from 5.00 in. opening to 500 mesh.

Compliance

Meet current ASTM E 11 and ISO 3310-1 specifications

Specifications

Specifications

Ideal for particle measurement in agricultural, bio industry, chemical, cosmetics, general industrial, fertilizer, foods, minerals, pharmaceuticals, Plastics and Metal products industries
8.2cm
4mm
ASTM E11, ISO 3310-1
Stainless Steel
Test sieve certificate
5 Mesh
Stainless Steel
3.25 in.
305mm
Full Height Test Sieve
12 in.
4.25 in.
10.7cm
No. 5
Documents
Accessories