missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ U.S. Standard Brass Sieves with Steel Cloth, 8 in. dia. × 1 in.D

Brass with stainless-steel cloth. Aperture sizes from 5.00 in. opening to 500 mesh.

Manufacturer:  Fisherbrand™ 5047*

 View more versions of this product

Catalog No. 04-881-42


 

Description

Description

Brass with stainless-steel cloth. Aperture sizes from 5.00 in. opening to 500 mesh.

Specifications

Specifications

Ideal for particle measurement in agricultural, bio industry, chemical, cosmetics, general industrial, fertilizer, foods, minerals, pharmaceuticals, Plastics and Metal products industries
2.5cm
75μm
ASTM E11, ISO 3310-1
Brass
Test sieve certificate
200 Mesh
Stainless Steel
1 in.
203mm
Half Height Test Sieve
8 in.
1.62 in.
4.11cm
No. 200
Documents