missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ U.S. Standard Brass Sieves with Steel Cloth, 8 in. dia. × 1 in.D

Brass with stainless-steel cloth. Aperture sizes from 5.00 in. opening to 500 mesh.

Manufacturer:  Fisherbrand™ 5043

 View more versions of this product

Catalog No. 04-881-41


 

Description

Description

Brass with stainless-steel cloth. Aperture sizes from 5.00 in. opening to 500 mesh.

Specifications

Specifications

Ideal for particle measurement in agricultural, bio industry, chemical, cosmetics, general industrial, fertilizer, foods, minerals, pharmaceuticals, Plastics and Metal products industries
2.5cm
150μm
ASTM E11, ISO 3310-1
Brass
Test sieve certificate
100 Mesh
Stainless Steel
1 in.
203mm
Half Height Test Sieve
8 in.
1.62 in.
4.11cm
No. 100
Documents
Accessories