Saint-Gobain Tygon S3™ E-3603 Flexible Tubings
Formulated for resistance to flex-fatigue and abrasion.
$47.36 - $1,104.03
Catalog Number | Mfr. No. | Diameter (English) Inner | Diameter (English) Outer | Thickness (English) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (English) Inner | Diameter (English) Outer | Thickness (English) | Price | Quantity | |||
14-171-208
|
Saint-Gobain
ACF00001F |
1/32 in. | 3/32 in. | 3/32 in. |
Pack of 50 Each for $59.03
|
|
14-171-129
|
Saint-Gobain
ACF00002F |
1/16 in. | 1/8 in. | 1/32 in. |
Pack of 50 Each for $52.48
|
|
14-171-130
|
Saint-Gobain
ACF00004F |
3/32 in. | 5/32 in. | 1/32 in. |
Pack of 50 Each for $47.36
|
|
14-171-211
|
Saint-Gobain
ACF00006F |
1/8 in. | 3/16 in. | 1/32 in. |
Pack of 50 Each for $49.33
|
|
14-171-215
|
Saint-Gobain
ACF00011F |
3/16 in. | 1/4 in. | 1/32 in. |
Pack of 50 Each for $76.15
|
|
14-171-131
|
Saint-Gobain
ACF00014F |
3/16 in. | 7/16 in. | 1/8 in. |
Pack of 50 Each for $401.00
|
|
14-171-104
![]() |
Saint-Gobain
ACF00017F |
1/4 in. | 3/8 in. | 1/16 in. |
Pack of 50 Each for $188.37
|
![]() |
14-171-132
|
Saint-Gobain
ACF00018F |
1/4 in. | 7/16 in. | 3/32 in. |
Pack of 50 Each for $321.18
|
|
14-171-221
|
Saint-Gobain
ACF00019F |
1/4 in. | 1/2 in. | 1/8 in. |
Pack of 50 Each for $388.64
|
|
14-171-223
|
Saint-Gobain
ACF00022F |
5/16 in. | 7/16 in. | 1/16 in. |
Pack of 50 Each for $230.09
|
|
14-171-224
|
Saint-Gobain
ACF00023F |
5/16 in. | 1/2 in. | 3/32 in. |
Pack of 50 Each for $373.39
|
|
14-171-227
|
Saint-Gobain
ACF00027F |
3/8 in. | 1/2 in. | 1/16 in. |
Pack of 50 Each for $297.09
|
|
14-171-228
|
Saint-Gobain
ACF00028F |
3/8 in. | 9/16 in. | 3/32 in. |
Pack of 50 Each for $384.06
|
|
14-171-133
|
Saint-Gobain
ACF00029F |
3/8 in. | 5/8 in. | 1/8 in. |
Pack of 50 Each for $415.94
|
|
14-171-234
|
Saint-Gobain
ACF00036F |
1/2 in. | 5/8 in. | 1/16 in. |
Pack of 50 Each for $361.48
|
|
14-171-235
|
Saint-Gobain
ACF00037F |
1/2 in. | 11/16 in. | 3/32 in. |
Pack of 50 Each for $502.06
|
|
14-171-236
|
Saint-Gobain
ACF00038F |
1/2 in. | 3/4 in. | 1/8 in. |
Pack of 50 Each for $761.53
|
|
14-171-242
|
Saint-Gobain
ACF00046F |
5/8 in. | 7/8 in. | 1/8 in. |
Pack of 50 Each for $781.18
|
|
14-171-245
|
Saint-Gobain
ACF00053F |
3/4 in. | 1 in. | 1/8 in. |
Pack of 50 Each for $978.80
|
|
14-171-252
|
Saint-Gobain
ACF00062F |
1 in. | 1-1/4 in. | 3/8 in. |
Pack of 50 Each for $1,104.03
|
|
Description
Formulated for resistance to flex-fatigue and abrasion.
- Bio-based formulation and contain no BPA or phthalates
- Provide high chemical resistance
- Non-oxidizing and non-contaminating
- Smooth, polished inner wall helps prevent buildup to facilitate cleaning
- Coils are marked at 30.4cm (1 ft.) intervals for easy measuring
- Slips easily over fittings and grips securely for simple lab set-ups
- Extra-heavy walls withstand full vacuum at room temperature (759mm [29.9 in.] of mercury at 23°C [73°F] and up to 686mm [27 in.] of mercury at 60°C [140°F])
- Sterilized by conventional autoclave methods (steam 30 min. at 15psi, 121°C) and Ethylene Oxide
Applications:
- Instrumentation connection, vent, drain and other general laboratory applications
- Food and beverage
- Biopharmaceutical
- Analytical instruments
- Peristaltic and vacuum pumps
- Condensers
- Incubators
- Desiccators
- Gas lines
- Drain lines
Compliance
FDA, NSF, 3-A, meets USP Class VI, Japan Food Sanitation Law no. 370/1959, REACH, 1935/2004/EC and 10/2011/EU.
Specifications
50 ft. | |
15 m |