Saint-Gobain Tygon S3™ E-3603 Flexible Tubings
Formulated for resistance to flex-fatigue and abrasion.
Supplier: Saint-Gobain ACF00046F
Description
Formulated for resistance to flex-fatigue and abrasion.
- Bio-based formulation and contain no BPA or phthalates
- Provide high chemical resistance
- Non-oxidizing and non-contaminating
- Smooth, polished inner wall helps prevent buildup to facilitate cleaning
- Coils are marked at 30.4cm (1 ft.) intervals for easy measuring
- Slips easily over fittings and grips securely for simple lab set-ups
- Extra-heavy walls withstand full vacuum at room temperature (759mm [29.9 in.] of mercury at 23°C [73°F] and up to 686mm [27 in.] of mercury at 60°C [140°F])
- Sterilized by conventional autoclave methods (steam 30 min. at 15psi, 121°C) and Ethylene Oxide
Applications:
- Instrumentation connection, vent, drain and other general laboratory applications
- Food and beverage
- Biopharmaceutical
- Analytical instruments
- Peristaltic and vacuum pumps
- Condensers
- Incubators
- Desiccators
- Gas lines
- Drain lines
Specifications
5/8 in. | |
50 ft. | |
1/8 in. |
7/8 in. | |
15 m | |
Clear |