missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Two-Way Slant Racks

For incubating liquid cultures on slant during tissue culture processes.

Manufacturer:  Fisherbrand™ 187873000

 View more versions of this product

Catalog No. 14-796-1




Description

Description

For incubating liquid cultures on slant during tissue culture processes.

  • Prepare either 5° or 20° agar slants in same rack
  • Autoclave media in rack
  • While still hot, set rack on one side for 5° tilt, or other for 20° tilt
  • 5° angle usually used for aerobic cultures, 20° for anaerobic
  • 20° tilt also prevents media from drying out during long-term storage
  • White, epoxy-coated, heavy-gauge steel

Specifications

Specifications

Autoclavable
16 mm
2.5 in.
13 to 16mm
20 cm
40
8 cm
White
7.75 x 3.25 x 2.5 in. (20 x 8 x 6cm)
6 cm
7.75 in.
Epoxy Coated Heavy-gauge Steel Wire
3.25 in.
Tube Rack
Documents