missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Traceable™ Vaccine Refrigerator/Freezer Thermometer

Monitors temperature of vaccine in freezers, water baths, heating blocks, incubators, and refrigerators with enclosed temperature-buffered sensor

Manufacturer:  Fisherbrand™ CON4527

 View more versions of this product

Catalog No. 15-077-720



Description

Description

Monitors temperature of vaccine in freezers, water baths, heating blocks, incubators, and refrigerators with enclosed temperature-buffered sensor. Fisherbrand™ Traceable™ Vaccine Refrigerator/Freezer Thermometer includes a probe which is sealed in glycol solution and insulates sensor when refrigerator door is opened.

  • Triple display simultaneously shows Min/max and current temperatures, °F/°C switchable
  • Bottle insulates sensor from rapid temperature changes when refrigerator door is opened
  • 15-077-720 has a 5mL bottle designed for vaccines; 06-664-11 has a 10mL bottle
  • Glass bottle filled with glycol mimics vaccine temperature and fits in trays
  • Probe is sealed in glycol solution which insulates sensor when refridgerator door is opened
  • Ten-foot micro-cable permits refrigerator doors to close on it
  • Minimum/maximum memory monitors high and low readings overnight, on weekends, or for any time period
  • Alarm is programmable in 1° increments, sounds continuously when temperature rises above/falls below set points and can be turned off manually
  • High-impact, chemical-resistant ABS plastic case
  • Temperature range: -50 to +70°C/-58 to +158°F
  • Resolution: 1° accuracy: ±1C

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It provides traceability to standards provided by NIST.

Specifications

Specifications

−58° to 158°F
158°F
−58°F
Glycol
ISO 17025, A2LA, NIST
2.75 x 4.25 x 0.75 in. (7 x 10.7 x 1.9cm)
0.75 in.
Traceable certificate, flip open stand, wall mounting, Velcro™ and magnetic strips
1.25 in.
30.4cm
ABS Plastic
1°C
110g
10.7cm
Digital LCD
−50° to 70°C
70°C
−50°C
5mL
0.3 in.
Monitoring temperature in freezers, water baths, heating blocks, incubators and refrigerators
19mm
120 in.
70mm
3.17cm
Refrigerator/Freezer Thermometer with 5mL bottle
4 oz.
4.25 in.
±1°C
2.75 in.
Documents