missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Traceable™ One-Shot Conductivity Standards

Eliminates contamination with calibration within disposable container. Fisherbrand™ Traceable™ One-Shot Conductivity Standards are 100% compatible with all conductivity meters.

Manufacturer:  Fisherbrand™ 9876948

 View more versions of this product

Catalog No. 06-664-275



Description

Description

Eliminates contamination with calibration within disposable container. Fisherbrand™ Traceable™ One-Shot Conductivity Standards are 100% compatible with all conductivity meters.

  • Nominal Value : 150000 Micro (mhos/siemens)
  • Certified Reference Material
  • Accuracy: certificate stated uncertainty, or ±0.25 microsiemen or ±0.25%
  • Includes: 6-pack 100mL container, step-by-step calibration instructions, individual temperature compensation charts, traceability information, Traceable Certificate
Specifications

Specifications

Calibration and storage of conductivity meters/ probes
0.01 to 1.6%
200000 Microsiemens/Micromhos
[Cl-].[K+]
74.548
CHEBI:32588
6 x 100mL
200000 Microsiemens/Micromhos
7447-40-7
ClK
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium;chloride
4873
Yes
Potassium chloride: 0.01 to 13.5%

Chemical Identifiers

7447-40-7
74.548
4873
potassium;chloride
ClK
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
CHEBI:32588
[Cl-].[K+]
SDS
Documents