Fisherbrand™ Traceable™ Memory Hygrometer/Thermometer
Simultaneously displays both temperature and relative humidity on dual 1.12 in. high LCD.
Manufacturer:  Fisherbrand™ 4395
 View more versions of this product
Catalog No. 06-664-273
Description
Description
Simultaneously displays both temperature and relative humidity on dual 1.12 in. high LCD.
  • Minimum/maximum humidity and temperature readings
  • Auto-off function
  • w/Computer output
  • w/Programmable audible alarm
Specifications
Specifications
Programmable Audible Alarm
Used for memory hygrometer/thermometer
Calibration instructions
75% Calibration Standard
Documents