Fisherbrand™ Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM), NIST tested
Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.
$55.38 - $247.04
Includes: Each bottle is supplied with step-by-step calibration instructions, individual temperature compensation chart, traceability information, and Traceable Certificate.
Products 18
Catalog Number Mfr. No. Description Conductivity Price Quantity  
Catalog Number Mfr. No. Description Conductivity Price Quantity  
06-664-24
View Documents
Fisherbrand
9001195
NIST batch tested; Resistivity: 0.2megohm 5μS
Each for $149.60
09-328-2 Edge
View Documents
Fisherbrand
9001118
NIST batch tested; Resistivity: 0.01megohm 100μS
Each for $55.83
Edge
09-328-11
View Documents
Fisherbrand
9001172
NIST batch tested; Resistivity: 0.00071megohm 1413μS
Each for $55.38
09-328-3 Edge
View Documents
Fisherbrand
9001123
NIST batch tested; Resistivity: 0.001megohm 1000μS
Each for $55.83
Edge
09-328-5
View Documents
Fisherbrand
9876919
NIST batch tested; Resistivity: 0.00001megohm 100000μS
Each for $55.72
09-328-4 Edge
View Documents
Fisherbrand
9001127
NIST batch tested; Resistivity: 0.0001megohm 10000μS
Each for $55.83
Edge
15-077-946
View Documents
Fisherbrand
9876946
NIST Individual bottle tested; Resistivity: 1.0megohm 1μS
Each for $246.57
15-077-947
View Documents
Fisherbrand
9876944
NIST Individual bottle tested; Resistivity: 0.5megohm 5μS
Each for $247.04
15-077-948
View Documents
Fisherbrand
9876939
NIST Individual bottle tested; Resistivity: 0.1megohm 10μS
Each for $244.46
15-077-952
View Documents
Fisherbrand
9876942
NIST Individual bottle tested; Resistivity: 0.0001megohm 10000μS
Each for $244.46
15-077-950
View Documents
Fisherbrand
9876941
NIST Individual bottle tested; Resistivity: 0.001megohm 1000μS
Each for $244.46
15-077-953
View Documents
Fisherbrand
9876943
NIST Individual bottle tested; Resistivity: 0.00001megohm 100000μS
Each for $244.94
15-077-949
View Documents
Fisherbrand
9876940
NIST Individual bottle tested; Resistivity: 0.01megohm 100μS
Each for $244.46
15-077-951
View Documents
Fisherbrand
9876945
NIST Individual bottle tested; Resistivity: 0.00071megohm 1413μS
Each for $244.46
06-664-265
View Documents
Fisherbrand
9876920
NIST batch tested; Resistivity: 150000μohms 150000μS
Each for $62.83
06-664-267
View Documents
Fisherbrand
9876937
NIST Individual bottle tested; Resistivity: 0.000006μohms 150000μS
Each for $187.38
06-664-266
View Documents
Fisherbrand
9876921
NIST batch tested; Resistivity: 200000μohms 200000μS
Each for $62.11
06-664-274
View Documents
Fisherbrand
9876938
NIST Individual bottle tested; Resistivity: 0.000005μohms 200000μS
Each for $189.13
Description
Description
Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.
Each bottle includes a NIST-traceable calibration report supplied by the manufacturer and information from batch testing. Also includes step-by step calibration instructions and temperature compensation chart. Accuracy at 25°C is (±0.25 microsiemens for 1, 5, 10 microsiemen solution or ±0.25% for other solutions) or the uncertainty shown on the certificate, whichever is greater.
Order Info
In 473mL (16 oz.) bottle