Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM), NIST tested, Fisherbrand™
Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.
Supplier:  Control Company 9876943
 View more versions of this product
Catalog No. 15-077-953
Description
Description
Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.
  • Each bottle includes a NIST-traceable calibration report supplied by the manufacturer and information from batch testing
  • Also includes step-by step calibration instructions and temperature compensation chart
  • Accuracy at 25°C is (±0.25 microsiemens for 1, 5, 10 microsiemen solution or ±0.25% for other solutions) or the uncertainty shown on the certificate, whichever is greater
  • ISO 17034 (1750.02) (formerly ISO Guide 34) Certified Reference Material
Order Info
473 mL (16 oz.) Bottle
Specifications
Specifications
NIST Individual Bottle Tested, Resistivity: 0.00001 mΩ
1 x 473 mL (16 oz.)
Clear
Conductivity Standard
ISO 17034 (1750.02) (formerly ISO Guide 34) Certified Reference Material
Bottle
Calibration Solution
NIST Individual Bottle Tested
100,000 μS
Liquid