Fisherbrand™ Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM), NIST tested
Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.
Manufacturer:  Fisherbrand™ 9876937
 View more versions of this product
Catalog No. 06-664-267
Description
Description
Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.
Each bottle includes a NIST-traceable calibration report supplied by the manufacturer and information from batch testing. Also includes step-by step calibration instructions and temperature compensation chart. Accuracy at 25°C is (±0.25 microsiemens for 1, 5, 10 microsiemen solution or ±0.25% for other solutions) or the uncertainty shown on the certificate, whichever is greater.
Specifications
Specifications
Calibration Solution
Conductivity Standard
150000μS
NIST Individual bottle tested; Resistivity: 0.000006μohms