Fisherbrand™ Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM), NIST tested
Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.
Manufacturer: Fisherbrand™ 9876921
Description
Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.
Each bottle includes a NIST-traceable calibration report supplied by the manufacturer and information from batch testing. Also includes step-by step calibration instructions and temperature compensation chart. Accuracy at 25°C is (±0.25 microsiemens for 1, 5, 10 microsiemen solution or ±0.25% for other solutions) or the uncertainty shown on the certificate, whichever is greater.Specifications
Calibration Solution | |
500mL | |
NIST batch tested; Resistivity: 200000μohms |
Conductivity Standard | |
200000μS |