Fisher Scientific™ Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM)
Use NIST-traceable reference materials for the highest accuracy available. Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.
Manufacturer:  Fisher Scientific™ CON4067
 View more versions of this product
Catalog No. 09-328-3
Specifications
Description
Standard, Calibration; Fisherbrand; Traceable; Conductivity; Dissolved solids: 666ppm; Resistivity: 0.001megohm; Conductivity: 1000μmho; 473mL (16 oz.); NIST batch tested
Each bottle includes a NIST-traceable calibration report supplied by the manufacturer and information from batch testing. Also includes step-by step calibration instructions and temperature compensation chart. Accuracy at 25°C is (±0.25 microsiemens for 1, 5, 10 microsiemen solution or ±0.25% for other solutions) or the uncertainty shown on the certificate, whichever is greater.
Description & Specifications
Specifications
16 oz. (475mL)
1000μS
NIST batch tested; Resistivity: 0.001megohm