Thermo Scientific™ Tissue Path™ II Cassettes
Standard lids fit securely onto cassettes
Manufacturer:  Thermo Scientific™ 58134
 View more versions of this product
Catalog No. 15-120-400J
Description
Description
  • Molded from a special high-density polymer resistant to all commonly used histological solvents
  • Slanted writing surface accepts markings easily, permitting positive sample identification
Order Info
Packaged in three dispenser boxes of 500
Specifications
Specifications
Separate Lid
Aqua
High Density Polymer
Tissue Cassette
Documents