Fisherbrand™ Tight-Seal™ Clamps
One-piece, reusable nylon clamps for hoses and tubing
Manufacturer:  Fisherbrand™ 9002438
 View more versions of this product
Catalog No. 05-815-1C
Description
Description
One-piece, reusable nylon clamps for hoses and tubing
  • Chemical and corrosion resistant
  • With wide temperature tolerance
  • Squeeze shut for leakproof seal, press on side to release
  • Gas or chemically sterilized for biomedical or food applications
  • 12 sizes accommodate tubing with an O.D. of 0.246 to 1.425 in. (6.25 to 36.19mm)
Pressure, vacuum, and liquid applications
Specifications
Specifications
0.41 to 0.46 in.
0°C to 90°C
Hose and tubing with 0.24 to 1.42 in O.D.
Hose and Tubing Clamp
32°F to 194°F
1.04 to 1.16 cm
Nylon