Fisherbrand™ Tight-Seal™ Clamps
One-piece, reusable nylon clamps for hoses and tubing
Manufacturer:  Fisherbrand™ 9002437
 View more versions of this product
Catalog No. 05-815-1D
Description
Description
One-piece, reusable nylon clamps for hoses and tubing
  • Chemical and corrosion resistant
  • With wide temperature tolerance
  • Squeeze shut for leakproof seal, press on side to release
  • Gas or chemically sterilized for biomedical or food applications
  • 12 sizes accommodate tubing with an O.D. of 0.246 to 1.425 in. (6.25 to 36.19mm)
Pressure, vacuum, and liquid applications
Specifications
Specifications
Hose and Tubing Clamp
0.47 to 0.53 in.
32°F to 194°F
Hose and tubing with 0.24 to 1.42 in O.D.
Nylon
0°C to 90°C
1.193 to 1.34 cm