Please login to your online account to display your discounted pricing
Bradley™ Thermostatic Mixing Valve
To further protect your employees from injury, choose the Bradley Thermostatic Mixing Valve. This device tempers eye/facewash water to prevent scalding.
$920.61 - $920.61
Products
Catalog Number Mfr. No. Price Quantity    
191-202-318
bradley
S192000*
Each for $920.61
Description & Specifications
For use with emergency eyewash, drench showers, and combination units. Built-in cold water bypass ensures a supply of cold water, even if the hot water supply is lost.
Single valve design
Reliable liquid filled thermostat with 10-year warranty
Factory pre-set at 85°F
Adjustable temperature range of 65 to 95°F
Universal mounting capability
Positive shutoff of hot supply when cold supply is lost
Built-in cold water bypass assures cold flow
Easy installation and serviceability
Dial thermometer
Rough bronze or chrome plated finish
It is the responsibility of the installer to determine the delivered water temperature to each safety fixture. The high-temperature limit stop is merely a mechanical setting to prevent excessive handle rotation. If the temperature of incoming water exceeds 60°C (150°F), delivery water at full hot-valve setting may exceed 32°C (90°F) and limit stop must be reset by the installer.
Meets ANSI Standard Z358.1-2004.