missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ PTFE Starburst Stirring Heads Individual Heads

Provide better mixing at bottom of vessel with less splashing.

Manufacturer:  Fisherbrand™ 800371074

 View more versions of this product

Catalog No. 14-513-79




Description

Description

Provide better mixing at bottom of vessel with less splashing.

  • Shape creates a deep vortex
  • Provides quiet, stable operation
  • Alnico magnet encapsulated in FDA grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclavable

Specifications

Specifications

Yes
PTFE
1.5 in.
38.1mm
Alnico Magnet
ISO 9001:2008
White
Stir bar
38.1mm
Documents
Accessories