Fisherbrand™ Test Tube Racks with Biohazard Warning
Easily identified in the lab and sterilization area by bright orange epoxy coating.
$66.97 - $66.97
Catalog Number | Mfr. No. | Holds | Positions | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Holds | Positions | Price | Quantity | ||||
14-793-34E
|
Fisherbrand™
187723003 |
Tubes (20 to 25 mm) | 40 |
Each for $66.97
|
|
Description
- Designed for use with biologically hazardous materials
- Also applicable in research with viruses, cancerous cells and recombinant DNA
- Sterilization can be performed with tubes in racks prior to discarding hazardous materials
- Universal biohazard symbol tag attached to each rack
- Epoxy coated steel wire; chemical- and corrosion-resistant
- Will withstand a temperature range of -223°C (-369°F) to 121°C (250°F); autoclavable
- All racks have square holes, three decks, a slotted grid bottom and reinforced edges
Specifications
Test Tube Rack |