Please login to your online account to display your discounted pricing

Fisherbrand™ Syringe Filters: Nylon Membrane


Low hold-up volume helps you to recover virtually all of your sample after filtering

Manufacturer: fisherbrand™  SE4M036I09

 View more versions of this product

Catalog No. 09-719-009

  • / Pack of 1500 Each
 

Description & Specifications

Specifications

Hold up Volume (After Air Purge) Low
No. per Pack 1500
Sterile No
Pore Size 0.45μm
Type Syringe Filter
Quantity 1500/Pk.
Diameter (Metric) 33mm
Material Nylon (NYL)
Applications Ideal for aqueous/organic/HPLC applications