Fisherbrand™ Syringe Filters - Nonsterile, Nylon Membrane
Low hold-up volume helps you to recover virtually all of your sample after filtering
Manufacturer: Fisherbrand™ SE4M018I09
Description
Tested for 100% membrane and housing integrity, eliminating concerns about fluid loss.
Nylon Membrane
- Excellent chemical resistance and low protein binding
- Ideal for aqueous/biological application
- Hydrophilic
Specifications
Nylon (NYL) | |
Non-sterile | |
50/pk. |
0.2μm | |
33mm | |
Syringe Filter |