Fisherbrand™ Syringe Filters - Nonsterile, Nylon Membrane
Low hold-up volume helps you to recover virtually all of your sample after filtering
$312.97 - $312.97
Specifications
Diameter (Metric) 33mm
Type Syringe Filter
Material Nylon (NYL)
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Pore Size Price Quantity  
Catalog Number Mfr. No. Pore Size Price Quantity  
09-719-005 Fisherbrand™
SE4M018I09
0.2μm Pack of 50 Each for $312.97
Specifications
Description
Tested for 100% membrane and housing integrity, eliminating concerns about fluid loss.
Nylon Membrane
  • Excellent chemical resistance and low protein binding
  • Ideal for aqueous/biological application
  • Hydrophilic
Description & Specifications
Specifications
33mm
Syringe Filter
Nylon (NYL)