Fisherbrand™ Syringe Filters - Nonsterile, Durapore™ PVDF Membrane
Low hold-up volume helps you to recover virtually all of your sample after filtering
Description
Low hold-up volume helps you to recover virtually all of your sample after filtering
Tested for 100% membrane and housing integrity, eliminating concerns about fluid loss.
- Hydrophilic membrane binds far less protein than nitrocellulose, nylon or PTFE
- Ideal for filtration of protein-containing aqueous, mild organic solutions and other pharmaceutical/biological/biotech solutions
Specifications
Durapore Hydrophilic Polyvinylidene Fluoride (PVDF) | |
Non-sterile | |
Syringe Filter |
0.45 μm | |
1500/Pk. | |
33 mm |