Please login to your online account to display your discounted pricing
Fisherbrand™ Syringe Filters - Nonsterile, Durapore™ PVDF Membrane
Low hold-up volume helps you to recover virtually all of your sample after filtering
Manufacturer: fisherbrand™  SE4J040I09
 View more versions of this product
Catalog No. 09-719-003
  • / Pack of 150 Each
Description & Specifications
Specifications
Pore Size 0.45μm
Sterile No
Type Syringe Filter
Quantity 150/pk.
Diameter (Metric) 33mm
Material Durapore Hydrophilic Polyvinylidene Fluoride (PVDF)