Fisherbrand™ SureStack Long Reach Pipet Tips
Pipetting station with high quality tips
Manufacturer:  Fisherbrand™ 31962
 View more versions of this product
Catalog No. 02-100-504
Description
Description
  • Ideal for use with multi-channel pipetters, both 8 and 12 channel
  • Decks snap together and will not lift during pipetting
  • Recessed wells keep tips stable for mounting
  • Saves bench space
  • Lid can be adjusted to expose only 96 tips
  • Wide base design prevents stack from tipping
  • Trays manufactured from recycled plastic
  • Autoclaving not recommended: available presterilized
  • 10μL Long Reach tip features extended length (45mm) and graduation marks at 2, 5 and 10μL
  • 200μL pipet tip features a soft fit collar, graduation marks at 10, 50 and 100μL
Specifications
Specifications
Yes
200 μL
Yellow
Extended Length
Recycled Plastic
Non-sterile
Not Autoclavable